MAN998718055 Manchester Organics

2,4-Dibromo-6-fluorobenzoic acid

  • MDL number MFCD03094087
  • Molecular Weight : 297.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/K55900
Availability details for
Quantity
  • Availability
  • Add to Cart