MANH039FCDF4 Manchester Organics

2-Chloro-1,3-difluorobenzene

  • MDL number MFCD00142604
  • Molecular Weight : 148.538

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B56789
Availability details for
Quantity
  • Availability
  • Add to Cart