MANH039FCEA3 Manchester Organics

(R)-1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate

  • MDL number MFCD12547038
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/W19912
Availability details for
Quantity
  • Availability
  • Add to Cart