MANH039FCEB0 Manchester Organics

2-Bromo-1,3,5-trichlorobenzene

  • MDL number MFCD00060653
  • Molecular Weight : 260.343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C57227
Availability details for
Quantity
  • Availability
  • Add to Cart