MANH039FCEB2 Manchester Organics

4-Chloro-3,5-difluoropyridine hydrochloride

  • MDL number MFCD30749393
  • Cas Number : 2102409-72-1
  • Molecular Weight : 185.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D19447
Availability details for
Quantity
  • Availability
  • Add to Cart