MANH039FCEE2 Manchester Organics

3-Methoxy-4-methyl-phenol

  • MDL number MFCD16998731
  • Molecular Weight : 138.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F57483
Availability details for
Quantity
  • Availability
  • Add to Cart