MANH039FD11A Manchester Organics

6-Chloro-5-methoxypyridine-3-carbaldehyde

  • MDL number MFCD13189003
  • Cas Number : 1060801-67-3
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-5-methoxynicotinaldehyde
Inchi Code 1S/C7H6ClNO2/c1-11-6-2-5(4-10)3-9-7(6)8/h2-4H,1H3
Inchi Key SIUZYEAMJQXUQU-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/X56900
Availability details for
Quantity
  • Availability
  • Add to Cart