MANH039FD19F Manchester Organics

2-[2-Chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD16996251
  • Cas Number : 1146214-95-0
  • Molecular Weight : 306.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[2-chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C13H15BClF3O2/c1-11(2)12(3,4)20-14(19-11)9-7-8(13(16,17)18)5-6-10(9)15/h5-7H,1-4H3
Inchi Key AAJHNTLYZHVWSZ-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/X56854
Availability details for
Quantity
  • Availability
  • Add to Cart