MANH03BB2DB3 Manchester Organics

2-(4-Benzyloxy-2-methoxy-phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD24039500
  • Cas Number : 1626407-70-2
  • Molecular Weight : 340.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/E57942
Availability details for
Quantity
  • Availability
  • Add to Cart