MANH03D4E7E9 Manchester Organics

[(2S)-Morpholin-2-yl]methanol hydrochloride

  • MDL number MFCD13191722
  • Cas Number : 1313584-92-7
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M39572
Availability details for
Quantity
  • Availability
  • Add to Cart