MANH03D4E822 Manchester Organics

5-Bromo-3-fluoro-2-methylpyridine

  • MDL number MFCD12196979
  • Cas Number : 1162674-74-9
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-fluoro-2-methylpyridine
Inchi Code 1S/C6H5BrFN/c1-4-6(8)2-5(7)3-9-4/h2-3H,1H3
Inchi Key KQCSRDNKPIPYIS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/A58237
Availability details for
Quantity
  • Availability
  • Add to Cart