MANH049F1AE4 Manchester Organics

4,5,6,7-Tetrahydro-1H-indazole-3-carboxylic acid

  • MDL number MFCD02939914
  • Molecular Weight : 166.177

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L58569
Availability details for
Quantity
  • Availability
  • Add to Cart