MANH11E4FF5E Manchester Organics

4H-Furo[3,2-b]pyrrole-5-carboxylic acid

  • MDL number MFCD07368660
  • Molecular Weight : 151.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/H59647
Availability details for
Quantity
  • Availability
  • Add to Cart