MANH11E503B6 Manchester Organics

Lithium clavulanate

  • MDL number MFCD22666231
  • Molecular Weight : 205.09382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A59456

Safety Information

MSDS https://www.manchesterorganics.com/A59456
Availability details for
Quantity
  • Availability
  • Add to Cart