MANH11E50648 Manchester Organics

5-(Phenoxymethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00980479
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/C59504
Availability details for
Quantity
  • Availability
  • Add to Cart