MANH2F6230B5 Manchester Organics

2-[2-(Difluoromethyl)-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Molecular Weight : 322.079

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B96715
Availability details for
Quantity
  • Availability
  • Add to Cart