MANH30312AE0 Manchester Organics

[5-[(tert-Butoxycarbonylamino)methyl]-2-thienyl]boronic acid

  • MDL number MFCD11504845
  • Cas Number : 1072951-39-3
  • Molecular Weight : 257.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-{[(tert-butoxycarbonyl)amino]methyl}-2-thienylboronic acid
Inchi Code 1S/C10H16BNO4S/c1-10(2,3)16-9(13)12-6-7-4-5-8(17-7)11(14)15/h4-5,14-15H,6H2,1-3H3,(H,12,13)
Inchi Key LCYICDNCCZCYQI-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/Z61755
Availability details for
Quantity
  • Availability
  • Add to Cart