MANH30312B29 Manchester Organics

(2S,5R)-2,5-Dimethyl-morpholine

  • MDL number MFCD19237826
  • Molecular Weight : 115.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P61769
Availability details for
Quantity
  • Availability
  • Add to Cart