MANH50286A2B Manchester Organics

(2S)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD04115802
  • Cas Number : 1508261-86-6
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-methyl-2-pyrrolidinecarboxylic acid hydrochloride
Inchi Code 1S/C6H11NO2.ClH/c1-6(5(8)9)3-2-4-7-6;/h7H,2-4H2,1H3,(H,8,9);1H/t6-;/m0./s1
Inchi Key YWPABDPNGQMUFH-RGMNGODLSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S61932
Availability details for
Quantity
  • Availability
  • Add to Cart