MANH580D85D7 Manchester Organics

(8-Methoxy-2-methyl-5-quinolyl)boronic acid dihydrate

  • MDL number MFCD26959690
  • Molecular Weight : 253.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/V61498
Availability details for
Quantity
  • Availability
  • Add to Cart