MANH7F37A089 Manchester Organics

4,5-Dichlorophthalic acid

  • MDL number MFCD00002493
  • Molecular Weight : 235.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H62016
Availability details for
Quantity
  • Availability
  • Add to Cart