MANH7F37A08A Manchester Organics

1-Benzyloxy-3,5-difluorobenzene

  • MDL number MFCD18072457
  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A61963
Availability details for
Quantity
  • Availability
  • Add to Cart