MAT047021181 Matrix Scientific

Methyl 3,3,3-trifluoro-DL-lactate

  • MDL number MFCD02093316
  • Cas Number : 93496-85-6
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Purity 95
Country of Origin USA
Boiling point 60-64�mm
Product Link http://www.matrixscientific.com/002251.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002251.html
Availability details for
Quantity
  • Availability
  • Add to Cart