MAT047021189 Matrix Scientific

2-Chloro-3,5-dinitrobenzotrifluoride

  • MDL number MFCD00007076
  • Cas Number : 392-95-0
  • Molecular Weight : 270.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-59
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002259.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/002259.html
Availability details for
Quantity
  • Availability
  • Add to Cart