MAT047021195 Matrix Scientific

2-Chloro-3,6-difluorobenzyl alcohol

  • MDL number MFCD01631445
  • Cas Number : 261762-44-1
  • Molecular Weight : 178.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002265.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002265.html
Availability details for
Quantity
  • Availability
  • Add to Cart