MAT047021198 Matrix Scientific

2-Chloro-3,6-difluorobenzyl bromide

  • MDL number MFCD01631446
  • Cas Number : 90292-67-4
  • Molecular Weight : 241.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002267.html

Safety Information

hcodes Corrosive/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/002267.html
Availability details for
Quantity
  • Availability
  • Add to Cart