MAT047021199 Matrix Scientific

2-Chloro-3,6-difluorocinnamic acid

  • MDL number MFCD01631464
  • Cas Number : 261762-48-5
  • Molecular Weight : 218.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002268.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002268.html
Availability details for
Quantity
  • Availability
  • Add to Cart