MAT047021202 Matrix Scientific

2-Chloro-3,6-difluorophenylacetonitrile

  • MDL number MFCD01631447
  • Cas Number : 261762-54-3
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002271.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/002271.html
Availability details for
Quantity
  • Availability
  • Add to Cart