MAT047021211 Matrix Scientific

4-Fluoro-3-(trifluoromethyl)benzamide

  • MDL number MFCD00061269
  • Cas Number : 67515-57-5
  • Molecular Weight : 207.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-85
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002280.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002280.html
Availability details for
Quantity
  • Availability
  • Add to Cart