MAT047021241 Matrix Scientific

2-Bromo-2,3,3,3-tetrafluoropropionyl fluoride

  • MDL number MFCD00153724
  • Cas Number : 6129-62-0
  • Molecular Weight : 226.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 67
Product Link http://www.matrixscientific.com/001159.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/001159.html
Availability details for
Quantity
  • Availability
  • Add to Cart