MAT047021247 Matrix Scientific

2-Methyl-3-(trifluoromethyl)benzamide

  • MDL number MFCD01631495
  • Cas Number : 251651-26-0
  • Molecular Weight : 203.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002313.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002313.html
Availability details for
Quantity
  • Availability
  • Add to Cart