MAT047021281 Matrix Scientific

4-Ethoxy-3-methoxyphenylacetonitrile

  • MDL number MFCD00016402
  • Cas Number : 103796-52-7
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Purity 98
Country of Origin USA
Boiling point 156-158�4 Torr
Product Link http://www.matrixscientific.com/002365.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/002365.html
Availability details for
Quantity
  • Availability
  • Add to Cart