MAT047021288 Matrix Scientific

5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride

  • MDL number MFCD00173797
  • Cas Number : 166964-33-6
  • Molecular Weight : 281.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-148
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/002370.html

Safety Information

hcodes Corrosive/Moisture Sensitive
MSDS http://www.matrixscientific.com/002370.html
Availability details for
Quantity
  • Availability
  • Add to Cart