MAT047021311 Matrix Scientific

2-Methyl-3-nitrobenzotrifluoride

  • MDL number MFCD00042322
  • Cas Number : 6656-49-1
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 86�mm
Product Link http://www.matrixscientific.com/002391.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002391.html
Availability details for
Quantity
  • Availability
  • Add to Cart