MAT047021327 Matrix Scientific

2-Methyl-4,4,4-trifluorobutanol

  • MDL number MFCD00190639
  • Cas Number : 107103-95-7
  • Molecular Weight : 142.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Boiling point 134
Product Link http://www.matrixscientific.com/002410.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/002410.html
Availability details for
Quantity
  • Availability
  • Add to Cart