MAT047021337 Matrix Scientific

Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoro-propane

  • MDL number MFCD00317556
  • Cas Number : 69563-87-7
  • Molecular Weight : 578.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-160
Country of Origin USA
Product Link http://www.matrixscientific.com/002419.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002419.html
Availability details for
Quantity
  • Availability
  • Add to Cart