MAT047021345 Matrix Scientific

2-Fluoro-4-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD00070808
  • Cas Number : 197239-49-9
  • Molecular Weight : 194.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002426.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002426.html
Availability details for
Quantity
  • Availability
  • Add to Cart