MAT047021364 Matrix Scientific

6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine

  • MDL number MFCD00052901
  • Cas Number : 22123-09-7
  • Molecular Weight : 191.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Country of Origin USA
Product Link http://www.matrixscientific.com/002447.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002447.html
Availability details for
Quantity
  • Availability
  • Add to Cart