MAT047021365 Matrix Scientific

3-Chloro-4-(trifluoromethoxy)aniline

  • MDL number MFCD00190126
  • Cas Number : 64628-73-5
  • Molecular Weight : 211.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.454
Country of Origin USA
Product Link http://www.matrixscientific.com/002448.html

Safety Information

hcodes Toxic/Irritant/Refrigerate
MSDS http://www.matrixscientific.com/002448.html
Availability details for
Quantity
  • Availability
  • Add to Cart