MAT047021370 Matrix Scientific

2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride

  • MDL number MFCD00173789
  • Cas Number : 154934-99-3
  • Molecular Weight : 244.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158
Purity 97
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 54�15mm
Product Link http://www.matrixscientific.com/002465.html

Safety Information

hcodes Corrosive/Stored under Argon/Moisture Sensitive
MSDS http://www.matrixscientific.com/002465.html
Availability details for
Quantity
  • Availability
  • Add to Cart