MAT047021372 Matrix Scientific

2-Chloro-4-(trifluoromethyl)phenyl hydrazine

  • MDL number MFCD00052916
  • Cas Number : 86398-98-3
  • Molecular Weight : 210.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-41
Purity 97
Country of Origin USA
Boiling point 80-85�25mm
Product Link http://www.matrixscientific.com/002467.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002467.html
Availability details for
Quantity
  • Availability
  • Add to Cart