MAT047021374 Matrix Scientific

4-Bromo-3,3,4,4-tetrafluoro-1-butene

  • MDL number MFCD00042189
  • Cas Number : 18599-22-9
  • Molecular Weight : 206.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.627
Country of Origin USA
Boiling point 55
Product Link http://www.matrixscientific.com/001174.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001174.html
Availability details for
Quantity
  • Availability
  • Add to Cart