MAT047021381 Matrix Scientific

Ethyl 5-(4-chlorophenyl)-2-(trifluoromethyl)-3-furoate

  • MDL number MFCD00275556
  • Cas Number : 175276-59-2
  • Molecular Weight : 318.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-93
Country of Origin USA
Product Link http://www.matrixscientific.com/002476.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002476.html
Availability details for
Quantity
  • Availability
  • Add to Cart