MAT047021390 Matrix Scientific

5-Chloro-4-fluoro-2-nitroaniline

  • MDL number MFCD00052698
  • Cas Number : 104222-34-6
  • Molecular Weight : 190.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002486.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002486.html
Availability details for
Quantity
  • Availability
  • Add to Cart