MAT047021392 Matrix Scientific

3-Chloro-4-fluoro-DL-phenylglycine

  • MDL number MFCD01631397
  • Cas Number : 261762-99-6
  • Molecular Weight : 203.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/002489.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002489.html
Availability details for
Quantity
  • Availability
  • Add to Cart