MAT047021397 Matrix Scientific

4-Amino-2,3,5,6-tetrafluorobenzamide

  • MDL number MFCD00008000
  • Cas Number : 1548-74-9
  • Molecular Weight : 208.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001176.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001176.html
Availability details for
Quantity
  • Availability
  • Add to Cart