MAT047021399 Matrix Scientific

2-Chloro-4-fluorobenzamide

  • MDL number MFCD00042531
  • Cas Number : 88578-90-9
  • Molecular Weight : 173.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-149
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002494.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002494.html
Availability details for
Quantity
  • Availability
  • Add to Cart