MAT047021424 Matrix Scientific

3-Chloro-4-fluorophenylacetic acid

  • MDL number MFCD01631555
  • Cas Number : 705-79-3
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002518.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002518.html
Availability details for
Quantity
  • Availability
  • Add to Cart