MAT047021425 Matrix Scientific

3-Chloro-4-fluorophenylacetonitrile

  • MDL number MFCD01631553
  • Cas Number : 658-98-0
  • Molecular Weight : 169.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 130-134�Torr
Product Link http://www.matrixscientific.com/002519.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002519.html
Availability details for
Quantity
  • Availability
  • Add to Cart