MAT047021432 Matrix Scientific

3-Chloro-4-fluorotoluene

  • MDL number MFCD00079730
  • Cas Number : 1513-25-3
  • Molecular Weight : 144.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.18
Country of Origin USA
Boiling point 167-169
Product Link http://www.matrixscientific.com/002526.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002526.html
Availability details for
Quantity
  • Availability
  • Add to Cart